Anti-TLR10 Mouse Monoclonal Antibody [clone: 670719]

Supplier: R & D Systems
MAB6619
102886-560EA 566.89 USD
102886-560
Anti-TLR10 Mouse Monoclonal Antibody [clone: 670719]
Antibodies
Type: Primary
Antigen: TLR10
Clonality: Monoclonal
Clone: 670719
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR