Anti-STX12 Mouse Monoclonal Antibody [clone: 677201]

Supplier: R & D Systems
MAB6617
102886-556EA 577.02 USD
102886-556
Anti-STX12 Mouse Monoclonal Antibody [clone: 677201]
Antibodies
Type: Primary
Antigen: STX12
Clonality: Monoclonal
Clone: 677201
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR