Anti-UBE2K Mouse Monoclonal Antibody [clone: 701316]

Supplier: R & D Systems
MAB6609
102886-552EA 594.37 USD
102886-552
Anti-UBE2K Mouse Monoclonal Antibody [clone: 701316]
Antibodies
Type: Primary
Antigen: UBE2K
Clonality: Monoclonal
Clone: 701316
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR