Anti-ATG7 Mouse Monoclonal Antibody [clone: 683906]

Supplier: R & D Systems
MAB6608
102886-550EA 633.42 USD
102886-550
Anti-ATG7 Mouse Monoclonal Antibody [clone: 683906]
Antibodies
Simple Western

Type: Primary
Antigen: ATG7
Clonality: Monoclonal
Clone: 683906
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR