Anti-ARAF Mouse Monoclonal Antibody [clone: 684128]

Supplier: R & D Systems
MAB6607
102886-548EA 577.02 USD
102886-548
Anti-ARAF Mouse Monoclonal Antibody [clone: 684128]
Antibodies
Type: Primary
Antigen: ARAF
Clonality: Monoclonal
Clone: 684128
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR