Anti-NLRP10 Mouse Monoclonal Antibody [clone: 578502]

Supplier: R & D Systems
MAB6606
102886-546EA 594.37 USD
102886-546
Anti-NLRP10 Mouse Monoclonal Antibody [clone: 578502]
Antibodies
Type: Primary
Antigen: NLRP10
Clonality: Monoclonal
Clone: 578502
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR