Anti-RAB25 Mouse Monoclonal Antibody [clone: 467657]

Supplier: R & D Systems
MAB6605
102886-544EA 566.89 USD
102886-544
Anti-RAB25 Mouse Monoclonal Antibody [clone: 467657]
Antibodies
Type: Primary
Antigen: RAB25
Clonality: Monoclonal
Clone: 467657
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR