Anti-RTN4 Rat Monoclonal Antibody [clone: 523808]

Supplier: R & D Systems
MAB6596
102886-536EA 561.11 USD
102886-536
Anti-RTN4 Rat Monoclonal Antibody [clone: 523808]
Antibodies
Type: Primary
Antigen: RTN4
Clonality: Monoclonal
Clone: 523808
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR