Anti-GPRC5C Mouse Monoclonal Antibody [clone: 577315]

Supplier: R & D Systems
MAB6594
102886-532EA 594.37 USD
102886-532
Anti-GPRC5C Mouse Monoclonal Antibody [clone: 577315]
Antibodies
Type: Primary
Antigen Symbol: GPRC5C
Antigen Name: G protein-coupled receptor, class C, group 5, member C
Antigen Synonyms: RAIG-3, RAIG3
Clonality: Monoclonal
Clone: 577315
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR