Anti-DSG4 Mouse Monoclonal Antibody [clone: 683135]

Supplier: R & D Systems
MAB6589
102886-530EA 577.02 USD
102886-530
Anti-DSG4 Mouse Monoclonal Antibody [clone: 683135]
Antibodies
Type: Primary
Antigen: DSG4
Clonality: Monoclonal
Clone: 683135
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR