Anti-CD44 Mouse Monoclonal Antibody [clone: 740017]

Supplier: R & D Systems
MAB6577
102886-528EA 577.02 USD
102886-528
Anti-CD44 Mouse Monoclonal Antibody [clone: 740017]
Antibodies
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 740017
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR