Anti-CCND3 Mouse Monoclonal Antibody [clone: 588726]

Supplier: R & D Systems
MAB6570
102886-526EA 584.25 USD
102886-526
Anti-CCND3 Mouse Monoclonal Antibody [clone: 588726]
Antibodies
Type: Primary
Antigen: CCND3
Clonality: Monoclonal
Clone: 588726
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR