Anti-Vitamin D3 Mouse Monoclonal Antibody [clone: 685503]

Supplier: R & D Systems
MAB6566
102886-524EA 594.37 USD
102886-524
Anti-Vitamin D3 Mouse Monoclonal Antibody [clone: 685503]
Antibodies
Type: Primary
Antigen Symbol:
Antigen Name: Vitamin D3
Antigen Synonyms: Calcifediol, Calciol, Calcitriol, Cholecalciferol
Clonality: Monoclonal
Clone: 685503
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR