Anti-TSPAN9 Mouse Monoclonal Antibody [clone: 590409]

Supplier: R & D Systems
MAB6565
102886-522EA 566.89 USD
102886-522
Anti-TSPAN9 Mouse Monoclonal Antibody [clone: 590409]
Antibodies
Type: Primary
Antigen: TSPAN9
Clonality: Monoclonal
Clone: 590409
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR