Anti-MUP1 Mouse Monoclonal Antibody [clone: 663323]

Supplier: R & D Systems
MAB6560
102886-518EA 594.37 USD
102886-518
Anti-MUP1 Mouse Monoclonal Antibody [clone: 663323]
Antibodies
Type: Primary
Antigen Symbol: MUP1
Antigen Name: major urinary protein 1
Antigen Synonyms: Ltn-1, Lvtn-1, Mup-1, Mup-a, Mup10, Mup7, Up-1
Clonality: Monoclonal
Clone: 663323
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR