Anti-FSHR Mouse Monoclonal Antibody [clone: 626717]

Supplier: R & D Systems
MAB65591
102886-516EA 763.57 USD
102886-516
Anti-FSHR Mouse Monoclonal Antibody [clone: 626717]
Antibodies
Type: Primary
Antigen Symbol: FSHR
Antigen Name: follicle stimulating hormone receptor
Antigen Synonyms: FSHR1, FSHRO, LGR1, ODG1
Clonality: Monoclonal
Clone: 626717
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR