Anti-PPA1 Mouse Monoclonal Antibody [clone: 840941]

Supplier: R & D Systems
MAB6557
102886-512EA 577.02 USD
102886-512
Anti-PPA1 Mouse Monoclonal Antibody [clone: 840941]
Antibodies
Type: Primary
Antigen: PPA1
Clonality: Monoclonal
Clone: 840941
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR