Anti-KLRC3 Mouse Monoclonal Antibody [clone: 633810]

Supplier: R & D Systems
MAB6549
102886-510EA 594.37 USD
102886-510
Anti-KLRC3 Mouse Monoclonal Antibody [clone: 633810]
Antibodies
Type: Primary
Antigen: KLRC3
Clonality: Monoclonal
Clone: 633810
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR