Anti-SLC22A2 Mouse Monoclonal Antibody [clone: 640438]

Supplier: R & D Systems
MAB6547
102886-506EA 594.37 USD
102886-506
Anti-SLC22A2 Mouse Monoclonal Antibody [clone: 640438]
Antibodies
Type: Primary
Antigen: SLC22A2
Clonality: Monoclonal
Clone: 640438
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR