Anti-DOT1L Mouse Monoclonal Antibody [clone: 653613]

Supplier: R & D Systems
MAB6546
102886-504EA 577.02 USD
102886-504
Anti-DOT1L Mouse Monoclonal Antibody [clone: 653613]
Antibodies
Type: Primary
Antigen: DOT1L
Clonality: Monoclonal
Clone: 653613
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR