Anti-HTR4 Mouse Monoclonal Antibody [clone: 631769]

Supplier: R & D Systems
MAB6545
102886-502EA 577.02 USD
102886-502
Anti-HTR4 Mouse Monoclonal Antibody [clone: 631769]
Antibodies
Type: Primary
Antigen: HTR4
Clonality: Monoclonal
Clone: 631769
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR