Anti-IL4 Mouse Monoclonal Antibody [clone: 286120]

Supplier: R & D Systems
MAB6542
220021-399EA 577.02 USD
220021-399
Anti-IL4 Mouse Monoclonal Antibody [clone: 286120]
Antibodies
Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone: 286120
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR