Anti-IL4 Mouse Monoclonal Antibody [clone: 99613]

Supplier: R & D Systems
MAB6541
220021-398EA 763.57 USD
220021-398
Anti-IL4 Mouse Monoclonal Antibody [clone: 99613]
Antibodies
Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone: 99613
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR