Anti-TOP2A Mouse Monoclonal Antibody [clone: 680130]

Supplier: R & D Systems
MAB6540
102886-498EA 594.37 USD
102886-498
Anti-TOP2A Mouse Monoclonal Antibody [clone: 680130]
Antibodies
Type: Primary
Antigen: TOP2A
Clonality: Monoclonal
Clone: 680130
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR