Anti-EDNRA Mouse Monoclonal Antibody [clone: 485711]

Supplier: R & D Systems
MAB6538
102886-496EA 594.37 USD
102886-496
Anti-EDNRA Mouse Monoclonal Antibody [clone: 485711]
Antibodies
Type: Primary
Antigen: EDNRA
Clonality: Monoclonal
Clone: 485711
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR