Anti-C1QTNF9 Mouse Monoclonal Antibody [clone: 653533]

Supplier: R & D Systems
MAB6537
102886-494EA 577.02 USD
102886-494
Anti-C1QTNF9 Mouse Monoclonal Antibody [clone: 653533]
Antibodies
Type: Primary
Antigen: C1QTNF9
Clonality: Monoclonal
Clone: 653533
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR