Anti-PLXNA1 Mouse Monoclonal Antibody [clone: 659807]

Supplier: R & D Systems
MAB6536
102886-492EA 577.02 USD
102886-492
Anti-PLXNA1 Mouse Monoclonal Antibody [clone: 659807]
Antibodies
Type: Primary
Antigen: PLXNA1
Clonality: Monoclonal
Clone: 659807
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR