Anti-TETX Mouse Monoclonal Antibody [clone: 604023]

Supplier: R & D Systems
MAB6535
102886-490EA 594.37 USD
102886-490
Anti-TETX Mouse Monoclonal Antibody [clone: 604023]
Antibodies
Type: Primary
Antigen: TETX
Clonality: Monoclonal
Clone: 604023
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: C. tetani
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR