Anti-TSHR Mouse Monoclonal Antibody [clone: 484405]

Supplier: R & D Systems
MAB6534
102886-488EA 594.37 USD
102886-488
Anti-TSHR Mouse Monoclonal Antibody [clone: 484405]
Antibodies
Type: Primary
Antigen: TSHR
Clonality: Monoclonal
Clone: 484405
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR