Anti-SERPINB3 Mouse Monoclonal Antibody [clone: 886524]

Supplier: R & D Systems
MAB6528
102886-484EA 594.37 USD
102886-484
Anti-SERPINB3 Mouse Monoclonal Antibody [clone: 886524]
Antibodies
Type: Primary
Antigen: SERPINB3
Clonality: Monoclonal
Clone: 886524
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR