Anti-AMPH Mouse Monoclonal Antibody [clone: 694145]

Supplier: R & D Systems
MAB6527
102886-480EA 561.11 USD
102886-480
Anti-AMPH Mouse Monoclonal Antibody [clone: 694145]
Antibodies
Type: Primary
Antigen: AMPH
Clonality: Monoclonal
Clone: 694145
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR