Anti-GNMT Mouse Monoclonal Antibody [clone: 691305]

Supplier: R & D Systems
MAB6526
102886-478EA 566.89 USD
102886-478
Anti-GNMT Mouse Monoclonal Antibody [clone: 691305]
Antibodies
Type: Primary
Antigen Symbol: GNMT
Antigen Name: glycine N-methyltransferase
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 691305
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR