Anti-TXK Mouse Monoclonal Antibody [clone: 674801]

Supplier: R & D Systems
MAB6520
102886-474EA 594.37 USD
102886-474
Anti-TXK Mouse Monoclonal Antibody [clone: 674801]
Antibodies
Type: Primary
Antigen: TXK
Clonality: Monoclonal
Clone: 674801
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR