Anti-IL2 Mouse Monoclonal Antibody [clone: 100321]

Supplier: R & D Systems
MAB652
220021-394EA 763.57 USD
220021-394
Anti-IL2 Mouse Monoclonal Antibody [clone: 100321]
Antibodies
Type: Primary
Antigen: IL2
Clonality: Monoclonal
Clone: 100321
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR