Anti-TEC Mouse Monoclonal Antibody [clone: 663921]

Supplier: R & D Systems
MAB6519
102886-472EA 594.37 USD
102886-472
Anti-TEC Mouse Monoclonal Antibody [clone: 663921]
Antibodies
Type: Primary
Antigen: TEC
Clonality: Monoclonal
Clone: 663921
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR