Anti-CD34 Rat Monoclonal Antibody [clone: 700011]

Supplier: R & D Systems
MAB6518
102886-470EA 594.37 USD
102886-470
Anti-CD34 Rat Monoclonal Antibody [clone: 700011]
Antibodies
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 700011
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR