Anti-TRIM32 Mouse Monoclonal Antibody [clone: 717801]

Supplier: R & D Systems
MAB6515
102886-468EA 561.11 USD
102886-468
Anti-TRIM32 Mouse Monoclonal Antibody [clone: 717801]
Antibodies
Type: Primary
Antigen: TRIM32
Clonality: Monoclonal
Clone: 717801
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR