Anti-RAMP2 Rat Monoclonal Antibody [clone: 773816]

Supplier: R & D Systems
MAB6500
102886-466EA 594.37 USD
102886-466
Anti-RAMP2 Rat Monoclonal Antibody [clone: 773816]
Antibodies
Type: Primary
Antigen: RAMP2
Clonality: Monoclonal
Clone: 773816
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR