Anti-MDC1 Rat Monoclonal Antibody [clone: 398636]

Supplier: R & D Systems
MAB6497
102886-462EA 577.02 USD
102886-462
Anti-MDC1 Rat Monoclonal Antibody [clone: 398636]
Antibodies
Type: Primary
Antigen: MDC1
Clonality: Monoclonal
Clone: 398636
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR