Anti-BRIP1 Mouse Monoclonal Antibody [clone: 652747]

Supplier: R & D Systems
MAB6496
102886-460EA 584.25 USD
102886-460
Anti-BRIP1 Mouse Monoclonal Antibody [clone: 652747]
Antibodies
Type: Primary
Antigen: BRIP1
Clonality: Monoclonal
Clone: 652747
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR