Anti-RB1 Mouse Monoclonal Antibody [clone: 607121]

Supplier: R & D Systems
MAB6495
102886-458EA 594.37 USD
102886-458
Anti-RB1 Mouse Monoclonal Antibody [clone: 607121]
Antibodies
Simple Western

Type: Primary
Antigen: RB1
Clonality: Monoclonal
Clone: 607121
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR