Anti-UGT1A1 Mouse Monoclonal Antibody [clone: 856754]

Supplier: R & D Systems
MAB6490
102886-456EA 594.37 USD
102886-456
Anti-UGT1A1 Mouse Monoclonal Antibody [clone: 856754]
Antibodies
Simple Western

Type: Primary
Antigen: UGT1A1
Clonality: Monoclonal
Clone: 856754
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR