Anti-ACAN (ADAMTS5-cleaved) Mouse Monoclonal Antibody [clone: 725913]

Supplier: R & D Systems
MAB64892
102886-454EA 633.42 USD
102886-454
Anti-ACAN (ADAMTS5-cleaved) Mouse Monoclonal Antibody [clone: 725913]
Antibodies
Type: Primary
Antigen: ACAN (ADAMTS5-cleaved)
Clonality: Monoclonal
Clone: 725913
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR