Anti-ACAN (ADAMTS5-cleaved) Mouse Monoclonal Antibody [clone: 661725]

Supplier: R & D Systems
MAB6489
102886-450EA 566.89 USD
102886-450
Anti-ACAN (ADAMTS5-cleaved) Mouse Monoclonal Antibody [clone: 661725]
Antibodies
Type: Primary
Antigen: ACAN (ADAMTS5-cleaved)
Clonality: Monoclonal
Clone: 661725
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR