Anti-HPGDS Mouse Monoclonal Antibody [clone: 735301]

Supplier: R & D Systems
MAB6487
102886-448EA 594.37 USD
102886-448
Anti-HPGDS Mouse Monoclonal Antibody [clone: 735301]
Antibodies
Type: Primary
Antigen: HPGDS
Clonality: Monoclonal
Clone: 735301
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR