Anti-PNP Mouse Monoclonal Antibody [clone: 773511]

Supplier: R & D Systems
MAB6486
102886-446EA 577.02 USD
102886-446
Anti-PNP Mouse Monoclonal Antibody [clone: 773511]
Antibodies
Type: Primary
Antigen: PNP
Clonality: Monoclonal
Clone: 773511
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR