Anti-PILRA Mouse Monoclonal Antibody [clone: 462415]

Supplier: R & D Systems
MAB6484
102886-444EA 594.37 USD
102886-444
Anti-PILRA Mouse Monoclonal Antibody [clone: 462415]
Antibodies
Type: Primary
Antigen: PILRA
Clonality: Monoclonal
Clone: 462415
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR