Anti-SLC22A1 Mouse Monoclonal Antibody [clone: 609222]

Supplier: R & D Systems
MAB6469
102886-438EA 561.11 USD
102886-438
Anti-SLC22A1 Mouse Monoclonal Antibody [clone: 609222]
Antibodies
Type: Primary
Antigen: SLC22A1
Clonality: Monoclonal
Clone: 609222
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR