Anti-C5AR1 Rat Monoclonal Antibody [clone: 583837]

Supplier: R & D Systems
MAB6467
102886-436EA 594.37 USD
102886-436
Anti-C5AR1 Rat Monoclonal Antibody [clone: 583837]
Antibodies
Type: Primary
Antigen: C5AR1
Clonality: Monoclonal
Clone: 583837
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR