Anti-GLB1 Mouse Monoclonal Antibody [clone: 810226]

Supplier: R & D Systems
MAB6464
102886-434EA 633.42 USD
102886-434
Anti-GLB1 Mouse Monoclonal Antibody [clone: 810226]
Antibodies
Type: Primary
Antigen Symbol: GLB1
Antigen Name: galactosidase, beta 1
Antigen Synonyms: EBP, ELNR1, MPS4B
Clonality: Monoclonal
Clone: 810226
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR